Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Iron Buffer Solution, Reagents
Click to view available options
Quantity:
500 mL
4 L
Specifications
Specifications
| Chemical Name or Material | Buffer |
| CAS | 6131-90-4 |
| Color | Colorless |
| For Use With (Application) | Iron Analysis |
| pH | 7 to 10 |
| Physical Form | Liquid |
| Quantity | 4 L |
| Packaging | Poly Bottle |
| Sterility | Non-sterile |
| Solubility Information | Completely miscible |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction