Learn More
Strem, An Ascensus Company CAS 635680-56-7. 5g. Bis(NN-di-t-butylacetamidinato)iron (II) min. 98%. MFCD08705281
SDP
Supplier:  Strem, An Ascensus Company 2601455G
Encompass_Preferred
CAS 635680-56-7. 5g. Bis(NN-di-t-butylacetamidinato)iron (II) min. 98%. MFCD08705281. Molecular Weight 394.42. Molecular Formula C20H42N4Fe. Color/form dark gray solid Strem 26-0145. http//www.strem.com/catalog/v/26-0145/
Catalog No. 50-211-0785
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.