Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Iron-doped (5 At%) Hydroxyapatite Nanopowder

Catalog No. 91554810G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-554-810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-554-810G Supplier Sigma Aldrich Supplier No. 91554810G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hydroxyapatite has been regarded as one of the best host materials for metallic ions doping, such as Ti, Zn, Fe, and Cu ions. Metal ion doping helps regulate the properties of hydroxyapatites and can reduce the cytotoxicity from metal ions.

Specifications

Quantity 10 g
Linear Formula (Ca5.Fe0.25)(PO4)3(OH)
Synonym Ca5(PO4)3 OH; Fe-doped HAp; Theranostic nanoparticles
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.