Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(II) Phthalocyanine (purified by sublimation) 98.0+%, TCI America™
SDP

Catalog No. I07831G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07831G 1 g
I0783200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07831G Supplier TCI America Supplier No. I07831G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 132-16-1
Molecular Formula C32H16FeN8
Molecular Weight (g/mol) 568.38
MDL Number MFCD00015953
InChI Key MIINHRNQLVVCEW-UHFFFAOYSA-N
Synonym iron phthalocyanine, iron ii phthalocyanine, ferrous phthalocyanine, phthalocyanine iron ii, fepc, 29h,31h-phthalocyaninato 2--n29,n30,n31,n32 iron, iron, 29h,31h-phthalocyaninato 2--n29,n39,n31,n32-, sp-4-1, iron, 29h,31h-phthalocyaninato 2--kappan29,kappan30,kappan31,kappan32-, sp-4-1
PubChem CID 2735065
IUPAC Name λ²-iron(2+) 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19,21(38),22,24,26,28,30,32,34-nonadecaene-37,39-diide
SMILES [Fe++].[N-]1C2=NC3=NC(=NC4=C5C=CC=CC5=C([N-]4)N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31

Specifications

Color Purple
Quantity 1 g
Formula Weight 568.38
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Iron(II) Phthalocyanine (purified by sublimation)
EINECSNumber (5)-4130
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.