Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(III) 1,1,1-trifluoro 2,4-pentanedionate

Catalog No. AA3926506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3926506 5 g
AA3926514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3926506 Supplier Thermo Scientific Chemicals Supplier No. 03926506
Add to Cart

CAS: 14526-22-8 | C15H12F9FeO6 | 515.09 g/mol

It is used as an intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14526-22-8
Molecular Formula C15H12F9FeO6
Molecular Weight (g/mol) 515.09
MDL Number MFCD00017760
InChI Key PDLKINIVWBCDRU-UHFFFAOYSA-N
Synonym ferric iron 1,1,1-trifluoro-4-oxopent-2-en-2-olate
PubChem CID 71299627
SMILES [Fe+3].CC(=O)[CH-]C(=O)C(F)(F)F.CC(=O)[CH-]C(=O)C(F)(F)F.CC(=O)[CH-]C(=O)C(F)(F)F

Specifications

Melting Point 110°C to 112°C
Quantity 5 g
Solubility Information Insoluble in water.
IUPAC Name iron(3+);(Z)-1,1,1-trifluoro-4-oxopent-2-en-2-olate
Formula Weight 515.09
Physical Form Crystalline
Chemical Name or Material Iron(III) 1,1,1-trifluoro 2,4-pentanedionate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.