Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(III) acetylacetonate, 99+%

Catalog No. p-150259
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC119130250 25 g Glass bottle
AC119131000 100 g Glass bottle
AC119132500 250 g Glass bottle
AC119130010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC119130250 Supplier Thermo Scientific Chemicals Supplier No. 119130250
Only null left
Add to Cart

CAS: 14024-18-1 | C15H21FeO6 | 353.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14024-18-1
Molecular Formula C15H21FeO6
Molecular Weight (g/mol) 353.17
MDL Number MFCD00000020
InChI Key AQBLLJNPHDIAPN-LNTINUHCSA-K
Synonym 2, 4-Pentanedione, iron(III) derivative, Ferric acetylacetonate
IUPAC Name iron(3+) tris((2Z)-4-oxopent-2-en-2-olate)
SMILES [Fe+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 180.0°C to 183.0°C
Color Red
Infrared Spectrum Authentic
Assay Percent Range 99% min. (Iodometry)
Packaging Glass bottle
Quantity 25 g
Fieser 04,268; 06,304; 13,211
Solubility Information Solubility in water: 2g/L (20°C). Other solubilities: readily soluble in acetone and chloroform,sparingly soluble in ether,40.1g/kg alcohol (20°C),535.4g/kg benzene (20°C)
Formula Weight 353.17
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material Iron(III) acetylacetonate, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 237-853-5
RTECSNumber NO8960000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.