Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(Iii) Chloride Hexahydrate, ACS Reagent, Solstice

Catalog No. 6000830 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
6 x 500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-008-30 100 g
60-008-31 500 g
60-008-33 6 x 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-008-30 Supplier Solstice Supplier No. 236489100G
Add to Cart
Add to Cart

ACS Reagent, 97%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-77-1
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Quantity 100 g
Linear Formula FeCl3 · 6H2O
UN Number 3260
Formula Weight 270.3g/mol
Percent Purity 97%
Grade ACS Reagent
Vapor Pressure 1mmHg (194°C)
Chemical Name or Material Iron(Iii) Chloride Hexahydrate
Hazard Statement P280-P305 + P351 + P338
Precautionary Statement H290-H302-H315-H318
EINECSNumber 231-729-4
RTECSNumber NO5425000
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.