Learn More
Medchemexpress LLC Iron(III) meso-tetra(N-methyl-4-pyridyl)porphine pentachloride | 133314-07-5 | 99.8% | 10 MG

Supplier: Medchemexpress LLC HY12038010MG
FeTMPyP is an iron-porphyrin complex used in research as a peroxynitrite decomposition catalyst and scavenger. It is supplied as a high-purity solid for laboratory studies and is intended for research use only.
- Acts as a peroxynitrite decomposition catalyst and scavenger.
- High purity (99.8%).
- Molecular weight 909.92 g/mol; chemical formula C44H36FeN8 · 5Cl.
- Supplied as a solid suitable for biochemical and in vivo studies.
- Recommended storage: sealed, away from moisture, -20°C (in solvent: -80°C up to 6 months; -20°C up to 1 month).
- Available in small pack sizes for precise dosing in experiments.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.