Learn More
Strem, An Ascensus Company CAS# 7782-61-8. 250g. Iron(III) nitrate nonahydrate, 98+%. MFCD00149708
Supplier: Strem, An Ascensus Company 932614250G
CAS# 7782-61-8. 250g. Iron(III) nitrate nonahydrate, 98+%. MFCD00149708. Molecular Weight: 241.86 (404.02). Molecular Formula: Fe(NO3)3∙9H2O. Color/form: light purple xtl. Strem# 93-2614. http://www.strem.com/catalog/v/93-2614/
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.