Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(III) trifluoromethanesulfonate, tech. 90%

Catalog No. AAH6003406
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6003406 5 g
AAH60034MD 250 mg
AAH6003403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6003406 Supplier Thermo Scientific Chemicals Supplier No. H6003406
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 63295-48-7 | C3F9FeO9S3 | 503.035 g/mol

Iron(III) triflate is used as an efficient catalyst in the synthesis of 2,3-unsaturated-O-glycosides from 2,4,6-tri-O-acetyl-D-glucal. It is also involved in the synthesis of a variety of beta-enamino ketones and esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iron(III) triflate is used as an efficient catalyst in the synthesis of 2,3-unsaturated-O-glycosides from 2,4,6-tri-O-acetyl-D-glucal. It is also involved in the synthesis of a variety of beta-enamino ketones and esters.

Solubility
Soluble in water, methanol and acetonitrile.

Notes
Moisture sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63295-48-7
Molecular Formula C3F9FeO9S3
Molecular Weight (g/mol) 503.035
MDL Number MFCD15144785
InChI Key OSHOQERNFGVVRH-UHFFFAOYSA-K
Synonym iron iii trifluoromethanesulfonate, fe otf 3, ferric triflate, iron iii triflate, fe cf3so3 3, ksc497q0p, iron 3+ tritriflate, ferric trifluoromethanesulfonate, iron 1,1,1-trifluoromethanesulfonic acid, iron 3+ ion tritrifluoromethanesulfonate
PubChem CID 11656316
IUPAC Name iron(3+);trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Fe+3]

Specifications

Melting Point 183°C
Flash Point 110°C (230°F)
Linear Formula Fe(OSO2CF3)3
Quantity 5 g
Sensitivity Moisture sensitive
Solubility Information Soluble in water,methanol and acetonitrile.
Formula Weight 503.04
Percent Purity 90%
Grade Technical
Chemical Name or Material Iron(III) trifluoromethanesulfonate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.