Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron Nickel powder, -325 mesh, 99.5+% (metals basis)

Catalog No. p-4781944
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3668314 25 g
AA3668322 100 g
AA3668336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3668314 Supplier Thermo Scientific Chemicals Supplier No. 03668314
Only null left
Add to Cart

Ni:Fe; 50:50 wt%

Iron Nickel powder is used as an low expansion alloy for precision instruments, measuring tapes and weights

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color Silver to Gray
Chemical Name or Material Iron Nickel powder
Appearance Silver gray powder
Quantity 25 g
Mesh Size -325 Mesh
Odor Odorless
Form Powder
Assay Percent Range >99.5% (metals basis)
Molecular Formula Ni:Fe; 50:50 wt%
MDL Number MFCD01741928
Solubility Information Insoluble in water.
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.