Learn More
Sigma Organic Chemistry Iron oxide | 1EA0.999
Supplier: Sigma Organic Chemistry GF181492381EA
Iron oxide, 1EA
About This Item:
UNSPSC Code: 12352200
Storage: room temp
EINECS Number: 215-277-5
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.