Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron Standard, Supelco

Catalog No. 11100251 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-251 1 Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-251 Supplier MilliporeSigma Supelco Supplier No. 1.09972.0050
Add to Cart
Add to Cart

1000 mg Fe, (FeCl3 in 15% HCl) Titrisol™

  • Titrisol™ offers a wide range of concentrated single-element standards.
  • Dilute the concentrated solution to achieve the desired standard concentration.
  • Directly traceable to standard reference materials (SRM) from the National Institute of Standards and Technology (NIST).
  • Supplied in easy-to-handle plastic ampoules.
  • Accompanied with a batch-specific certificate of analysis (COA) containing information on the NIST batch and minimum shelf life.
  • Provides reliable and reproducible results.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Iron Standard
Quantity 1 Bottle
Form Liquid
Molecular Weight (g/mol) Unknown
Percent Purity Unknown
Grade Analytical
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.