Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Micronova™ Hazard Bags
BIOPHARMAGREENER_CHOICESDP

Catalog No. 19065142 Shop All Micronova Manufacturing Products
Change view
Click to view available options
Color:
Red
Yellow
Capacity (Metric):
60 L
61 L
125 L
Sterility:
Non-sterile
Sterile
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Capacity (Metric) Sterility
19-065-142 Red 60 L Sterile
19-263-623 Yellow 61 L Sterile
19-116-729 Red 125 L Sterile
19-116-737 Red 125 L Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-065-142 Supplier Micronova™ Supplier No. BL225REIRBB
Add to Cart
Add to Cart

Low density, LDPE hazard bags are designed for containing biomedical and hazardous waste in aseptic and controlled environments.

Micronova Hazard Bags are constructed from FDA compliant, low density, red or yellow polyethylene and can be used with the BLSSH series Micronova BinLiner Stands. The Biohazard bags are designed for containment and disposal of biomedical waste and the Hazardous Material bags are designed for segregation and disposal of hazardous waste

  • Low density polyethylene Hazard bags
  • Red or Yellow Biohazard and Hazardous Material bags
  • Biohazard bags are clearly marked with a Biohazard symbol in black ink
  • Hazardous material bags are labeled for waste segregation in the US
  • The 32YE liners work with the BLSSH-1515 BinLiner Stand
  • The 24YE and 25RE work with the BLSSH-168 BinLiner Stand
  • Smaller bags with a star bottom are ideal for round containers
  • Larger bags are flat sealed
  • Good compatibility with common cleaners
  • Gamma irradiated liners are sterile for aseptic areas

Specifications

Capacity (Metric) 60 L
Certifications/Compliance Certificate of Conformance and Certificate of Irradiation Processing; ISO 9001 Certified Facility Meets ASTM D170910-6 Sterility Assurance Level In-Compliance with AAMI Guidelines for Sterilization.
Description Five folded liners per double bag
Diameter (Metric) Outer 40 cm
Diameter (English) Outer 15.9 in.
Capacity (English) 16 gal.
Color Red
Dimensions (L x W) 25 x 35 in.
Disposable Single-use
Height (Metric) 91 cm
Length (Metric) 63.5 cm
Quantity 50/Cs.
Shape STAR: Durable pleated bottom to fit round containers.
Sterility Sterile
Autoclavable Not Autoclavable
For Use With (Application) Transporting Biomedical Waste (Non-Sharps)
For Use With (Equipment) BLSSH series BinStand or similar container
Length (English) 25 in.
Thickness 0.050 mm
Height (English) 36 in.
Material LDPE
Product Type Biohazard Waste Bag
Temperature (Metric) 15°C to 30°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.