Learn More
Description
Micronova Hazard Bags are constructed from FDA compliant, low density, red or yellow polyethylene and can be used with the BLSSH series Micronova BinLiner Stands. The Biohazard bags are designed for containment and disposal of biomedical waste and the Hazardous Material bags are designed for segregation and disposal of hazardous waste
- Low density polyethylene Hazard bags
- Red or Yellow Biohazard and Hazardous Material bags
- Biohazard bags are clearly marked with a Biohazard symbol in black ink
- Hazardous material bags are labeled for waste segregation in the US
- The 32YE liners work with the BLSSH-1515 BinLiner Stand
- The 24YE and 25RE work with the BLSSH-168 BinLiner Stand
- Smaller bags with a star bottom are ideal for round containers
- Larger bags are flat sealed
- Good compatibility with common cleaners
- Gamma irradiated liners are sterile for aseptic areas
Specifications
Specifications
| Capacity (Metric) | 125 L |
| Certifications/Compliance | Certificate of Conformance and Certificate of Irradiation Processing; ISO 9001 Certified Facility Meets ASTM D170910-6 Sterility Assurance Level In-Compliance with AAMI Guidelines for Sterilization. |
| Description | Five folded liners per double bag |
| Diameter (Metric) Outer | 55 cm |
| Diameter (English) Outer | 21.7 in. |
| Capacity (English) | 33 gal. |
| Color | Red |
| Dimensions (L x W) | 34 x 40 in. |
| Disposable | Single-use |
| Height (Metric) | 102 cm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.


