Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Micronova™ Hazard Bags
BIOPHARMAGREENER_CHOICESDP

Catalog No. 19116737 Shop All Micronova Manufacturing Products
Change view
Click to view available options
Color:
Red
Yellow
Capacity (Metric):
60 L
61 L
125 L
Sterility:
Non-sterile
Sterile
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Capacity (Metric) Sterility
19-116-737 Red 125 L Non-sterile
19-263-623 Yellow 61 L Sterile
19-065-142 Red 60 L Sterile
19-116-729 Red 125 L Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-116-737 Supplier Micronova™ Supplier No. BL23440REBB
Add to Cart
Add to Cart
This item is not returnable. View return policy

Low density, LDPE hazard bags are designed for containing biomedical and hazardous waste in aseptic and controlled environments.

Micronova Hazard Bags are constructed from FDA compliant, low density, red or yellow polyethylene and can be used with the BLSSH series Micronova BinLiner Stands. The Biohazard bags are designed for containment and disposal of biomedical waste and the Hazardous Material bags are designed for segregation and disposal of hazardous waste

  • Low density polyethylene Hazard bags
  • Red or Yellow Biohazard and Hazardous Material bags
  • Biohazard bags are clearly marked with a Biohazard symbol in black ink
  • Hazardous material bags are labeled for waste segregation in the US
  • The 32YE liners work with the BLSSH-1515 BinLiner Stand
  • The 24YE and 25RE work with the BLSSH-168 BinLiner Stand
  • Smaller bags with a star bottom are ideal for round containers
  • Larger bags are flat sealed
  • Good compatibility with common cleaners
  • Gamma irradiated liners are sterile for aseptic areas

Specifications

Capacity (Metric) 125 L
Certifications/Compliance ISO 9001 Certified Facility; Certificate of Conformance; FDA Regulation 21 CFR, 177.1520, 3.1
Description Five folded liners per double bag
Diameter (Metric) Outer 55 cm
Diameter (English) Outer 21.7 in.
Capacity (English) 33 gal.
Color Red
Disposable Single-use
Height (Metric) 102 cm
Length (Metric) 86 cm
Quantity 50/Cs.
Shape Star Bottom: durable pleated bottom to fit round containers.
Sterility Non-sterile
Autoclavable Not Autoclavable
For Use With (Application) Designed for the collection and containment of biomedical waste.
For Use With (Equipment) BLSSH-1515 or similar container
Length (English) 34 in.
Thickness 0.050 mm
Height (English) 40 in.
Material LDPE
Product Type Biohazard Waste Bag
Temperature (Metric) 15°C to 30°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.