Learn More
R&D Systems™ Irradiated Mouse Embryonic Fibroblasts (5 vials)
SDP
Catalog No. PSC001 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
6 x 10e6 Cells/Vial, 5 Vials
Catalog No. Quantity
PSC001 6 x 10e6 Cells/Vial, 5 Vials
1 options
This item is not returnable. View return policy
Catalog No. PSC001 Supplier R&D Systems™ Supplier No. PSC001
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
For use as a feeder layer when culturing embryonic or induced pluripotent stem cells
Mitotically inactive mouse embryonic fibroblasts (MEFs) support the growth and maintenance of stem cell pluripotency by secreting necessary soluble factors and by providing a substrate for stem cell attachment.
  • Ready to use
  • MEFs support undifferentiated growth of embryonic and induced pluripotent stem cells
  • Inactivated by gamma irradiation to avoid chemical contamination
  • Consistent feeder layer to reduce experimental variation
  • Free of microbial and mycoplasma contamination
Specifications
Quantity 6 x 10e6 Cells/Vial, 5 Vials