Learn More
Thermco Products Irreversible Freeze Indicator
Catalog No. 04500336
Change view
Click to view available options
Temperature Range (Metric):
-5°C
2°C
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature Range (Metric)
04-500-336 -5°C
04-500-048 2°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 04-500-336 Supplier Thermco Products Supplier No. F1023E
Add to Cart
Add to Cart
Self-adhesive labels / Irreversible color change - Green to Red / Self-activating / Storage: Room Temperature
  • The ATI Freeze Check Indicator is a cost-effective, self-activating solution for monitoring freeze-sensitive products in the supply chain.
  • Durable, low-profile design withstands up to 100 psi and adheres to various surfaces, making it ideal for pharmaceuticals, food, and industrial goods.
  • Features a green circle with a checkmark, the temperature-sensitive liquid turns opaque white when exposed to freezing temperatures, providing clear and irreversible evidence of exposure.
  • No pre-activation is needed, simply apply the self-adhesive label.
  • Free of glass or hazardous materials, simplifying disposal and ensuring safety.
  • Ideal for cold chain logistics, freeze check reduces risks and costs by offering reliable, at-a-glance freeze detection for critical shipments.
Specifications
Material Paper
Product Type Chemical Temperature Indicator
Temperature Range (Metric) -5°C