Learn More
Description
PA1-1058 detects a IRS-1 in mouse and human cells. PA1-1058 has been successfully used in Western blot procedures. By Western blot this antibody detects an ~170 kDa protein representing IRS-1 in 3T3 L1 cell extract. The PA1-1058 immunogen is a synthetic peptide corresponding to residues L(602) H T D D G Y M P M S P G V(615) of human IRS-1. The immunizing peptide is 92% conserved in rats, and mice. This peptide (Cat. # PEP-192) is available for use in neutralization and control experiments.
Specifications
Specifications
| Antigen | IRS1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1 mg/mL BSA and 0.05% sodium azide |
| Gene | IRS1 |
| Gene Accession No. | P35568, P35569 |
| Gene Alias | G972R; HIRS-1; Insulin receptor substrate 1; IRS1; IRS-1; IRS1IRM; pp185 |
| Gene Symbols | IRS1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
