Learn More
TARGETMOL CHEMICALS INC ISATIN-BETA-THIOSEMICARB 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T602725MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Isatin-(beta)-thiosemicarbazone is a potent inhibitor of herpes simplex virus (HSV). isatin-(beta)-thiosemicarbazone exhibits antiviral activity by inhibiting both HSV-1 and HSV-2. isatin-(beta)-thiosemicarbazone is Isatin-(beta)-thiosemicarbazone is an effective anti-poxvirus agent for the treatment of monkeypox orthopox and cowpox viruses. purity: 98%
Catalog No. 50-257-9630
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.