Learn More
Selleck Chemical LLC Isavuconazole 5mg 241479-67-4 BAL-4815, RO-0094815
SDP
Supplier:  Selleck Chemical LLC S37225MG
Encompass_Preferred
Isavuconazole (BAL-4815, RO-0094815) is a new extended-spectrum triazole with activity against yeasts, molds, and dimorphic fungi that inhibits cytochrome P450 (CYP)-dependent 14?-lanosterol demethylation, which is essential for fungal cell membrane ergosterol synthesis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8321
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.