Learn More
Medchemexpress LLC HY-100373 5mg Medchemexpress, Isavuconazonium (sulfate) CAS:946075-13-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1003735MG
Encompass_Preferred
Medchemexpress, HY-100373 5mg Isavuconazonium (sulfate) CAS:946075-13-4 Isavuconazonium sulfate (BAL8557-002), the prodrug of the active triazole Isavuconazole, is an orally active antifungal agent. Isavuconazonium sulfate is used for invasive aspergillosis and mucormycosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0583
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.