Learn More
Selleck Chemical LLC ISCK03 S2070-25mg
SDP
Supplier:  Selleck Chemical LLC S207025MG
Encompass_Preferred
ISCK03 (N-(4-imidazol-1-yl phenyl)sulfonamide) is a cell-permeable inhibitor of stem-cell factor (SCF)/c-kit signaling ISCK03 inhibits SCF-induced c-c-kit phosphorylation and downstream ERK phosphorylation
Catalog No. 50-311-0372
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.