Learn More
Abbott™ iScreen™ Dip Drug Cards
Catalog No. 22911030
Change view
Click to view available options
Detectable Analytes:
Buprenorphine (BUP)
Cocaine (COC)
Cocaine (COC), Cannibinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP)
Cocaine (COC), Cannibinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Phencyclidine (PCP), Benzodiazepines (BZO), Barbiturates (BAR)
Cocaine (COC), Cannibinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Phencyclidine (PCP), Benzodiazepines (BZO), Barbiturates (BAR), Methadone (MTD)
Cocaine (COC), Cannibinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Phencyclidine (PCP), Benzodiazepines (BZO), Barbiturates (BAR), Methadone (MTD), Methylenedioxymethamphetamine (MDMA), Oxycodone (OXY), Propoxyphene (PPX)
Cocaine (COC), Cannibinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Phencyclidine (PCP), Benzodiazepines (BZO), Barbiturates (BAR), Methadone (MTD), Tricyclic Antidepressants (TCA)
Oxycodone (OXY)
Tricyclic Antidepressants (TCA)
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes
22-911-030 Buprenorphine (BUP)
22-911-040 Cocaine (COC), Cannibinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Phencyclidine (PCP), Benzodiazepines (BZO), Barbiturates (BAR), Methadone (MTD), Methylenedioxymethamphetamine (MDMA), Oxycodone (OXY), Propoxyphene (PPX)
22-911-056 Cocaine (COC)
22-911-039 Cocaine (COC), Cannibinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Phencyclidine (PCP), Benzodiazepines (BZO), Barbiturates (BAR), Methadone (MTD), Tricyclic Antidepressants (TCA)
22-911-037 Cocaine (COC), Cannibinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Phencyclidine (PCP), Benzodiazepines (BZO), Barbiturates (BAR), Methadone (MTD)
22-911-033 Cocaine (COC), Cannibinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP)
22-911-029 Oxycodone (OXY)
22-911-031 Tricyclic Antidepressants (TCA)
22-911-036 Cocaine (COC), Cannibinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Phencyclidine (PCP), Benzodiazepines (BZO), Barbiturates (BAR)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 22-911-030 Supplier Abbott™ Supplier No. DBU114
Add to Cart
Add to Cart
This item is not returnable. View return policy
Testing for up to 12 drugs on one device
Detect the presence of drugs in minutes
  • Simple dip and read procedure
  • Room temperature storage
  • Laboratory accurate
  • 1-12 Drug configurations available
  • Results in minutes
  • 24 Month shelf life
Specifications
Detectable Analytes Buprenorphine (BUP)
Sample Type Urine
Type iScreen Urine Drug Screen
Quantity 25/Pk.
Test Time Results in minutes