Learn More
Abbott™ iScreen™ Dip Drug Cards
Description
Detect the presence of drugs in minutes
- Simple dip and read procedure
- Room temperature storage
- Laboratory accurate
- 1-12 Drug configurations available
- Results in minutes
- 24 Month shelf life
Specifications
Specifications
| Detectable Analytes | Cocaine (COC), Cannibinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Phencyclidine (PCP), Benzodiazepines (BZO), Barbiturates (BAR), Methadone (MTD) |
| Sample Type | Urine |
| Type | iScreen Urine Drug Screen |
| Quantity | 25/Pk. |
| Test Time | Results in minutes |