Learn More
Roche Diagnostics ISE CALIBRATOR DIRECT BIO HT
Supplier:  Roche Diagnostics 06664172001
Encompass
Calibrator solution for one-point calibration of the ISE sensors with every direct sample measurement.
Catalog No. 50-589-425
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.