Learn More
Roche Diagnostics ISE Calibrator Indirect Bio, 2 x 230 mL
Supplier: Roche Diagnostics 06439691001
Reference solution for one-point calibration of the ISE sensors with every indirect sample measurement. Calibrator reagent for the Cedex Bio Analyzer.