Learn More
Roche Diagnostics ISE SOLUTION 2 6 X9.5 ML
Supplier:  Roche Diagnostics 06664202001
Encompass
ISE Solution 2, 6 x 9.5 ml System reagent for the ISE unit of the Cedex Bio HT Analyzer Calibrator solution for two-point calibration of the ISE sensors during initialization of the ISE module. - Specification Na: 110 2 mmol/L K: 1.8 0.066 mmol/L pH (25C): 7.45 0.15 For use in quality control / manufacturing process only. 64-19-7
Catalog No. 50-589-427
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.