Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech ISF-1 Medium for Hybridoma, w: stable Glutamine, w: Insulin hum. rec., w: 2.438 g/L NaHCO3 (1000 ml)
SDP
Supplier:  ILEX LIFE SCIENCES LLC P04995968
Encompass_Preferred
ISF-1 Medium for Hybridoma, w: stable Glutamine, w: Insulin hum. rec., w: 2.438 g/L NaHCO3. QUANTITY: 1000 ml. MANUFACTURED BY: Pan-Biotech (Germany). STORAGE: 2 to 8 degrees Celsius. DESCRIPTION: a serum-free and defined culture medium for in vitro cultivation and expansion of hybridoma cell lines. High yield and final concentration of monoclonal antibodies (mAb) can be achieved. As this product is free of hydrolysates, and only contains traces of animal-derived components (less than 0.1% w/v) and proteins (less than 0.1% w/v), it is suitable for the production of mAb in higher scales. COMPOSITION: ISF-1 has been manufactured with stable Glutamine. It also contains a surfactant. Therefore no additional supplement for agitated suspension culture is needed.
Catalog No. 50-238-8611
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.