Learn More
AVANTOR MASTERFLEX (VWR LLC) Ismatec ISM947C IP-N Digital Peristaltic Pump, Low-Speed, 12-Channel, Click-N-Go Cartridge, 11.25 rpm; 115 VAC
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX7801121
Encompass
This multichannel programmable pump features planetary gears that drive eight stainless steel rollers in unison for smooth rotation and accurate liquid flow. The four-digit LED display shows flow rate or rpm when pump is running. Use the MAX button to instantly increase speed for priming or flushing without disturbing your programmed settings. Combine tubing sizes and formulations to suit your application. Clickngo cartridges feature automatic occlusion for simplified use, fewer tubing changes, and reproducible results. Adjustable occulsion cartridges are available separately. Install cartridges in one direction for low pulsation; in opposite direction for higher counter pressure. Pressure Lever Cartridges offer the user the opportunity to manually adjust the back pressure experienced by the tubing / pump combination.
Catalog No. NC1054646
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.