Learn More
AVANTOR MASTERFLEX (VWR LLC) Ismatec IPC-N ISM938D Digital Peristaltic Pump, Low-Speed, Click-'N-Go Cartridges, 16-Channel, 11.25 rpm; 115 VAC
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX7800130
Encompass
Flow rate of 0.002 to 44 mL/min per channel - flow rate depends on drive rpm and tubing sizePlanetary drive for less pulsation and longer tubing life. Control via keypad, computer, or remote controlSmooth rotation and accurate liquid flow provided by eight SS rotors. Easy repeatability with four-digit LED. Simple to useautomatic occlusion cartridges provide reproducible results.
Catalog No. NC1767757
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.