Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ISO 1
GREENER_CHOICE

Catalog No. 428810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-881-0 10 mg
42-885-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 42-881-0 Supplier Tocris Bioscience™ Supplier No. 4288/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Macrophage migration inhibitory factor (MIF) inhibitor

ISO 1 is a macrophage migration inhibitory factor (MIF) inhibitor (IC50 = 7 μM); inhibits MIF tautomerase activity in vitro and in vivo. Protective against mouse models of streptozotocin -induced diabetes mellitus; blocks anti-inflammatory response following LPS exposure and increases survival. Inhibits airway remodeling in a mouse model of chronic asthma. Cell permeable; orally bioavailable.

Chemical Identifiers

CAS 478336-92-4
Molecular Formula C12H13NO4

Specifications

Quantity 10 mg
Formula Weight 235.24
Physical Form Solid
Color White
Purity >98%
Molecular Weight (g/mol) 235.239
Chemical Name or Material ISO 1
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.