Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

iso-Amyl Alcohol ACS, MilliporeSigma™

Catalog No. MAX14406 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MAX14406 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAX14406 Supplier MilliporeSigma Supplier No. US1AX1440/6
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 123-51-3
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
InChI Key PHTQWCKDNZKARW-UHFFFAOYSA-N
Synonym 3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
PubChem CID 31260
ChEBI CHEBI:15837
IUPAC Name 3-methylbutan-1-ol
SMILES CC(C)CCO

Specifications

Density 0.81g/cm3 (20°C)
Explosion Limit 1.2 to 8%(V)
Ignition Point 335°C
Packaging Glass Bottle
Linear Formula (CH3)2CHCH2CH2OH
Quantity 500 mL
Vapor Pressure 3 hPa (20°C)
Formula Weight 88.15
Grade ACS
Chemical Name or Material iso-Amyl Alcohol, For Synthesis
Recommended Storage Store at +15°C to +25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.