Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iso-Butyl Alcohol, ACS Reagent (2-Methyl-1-Propanol), Reagents

Catalog No. 01338148
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Glass Bottle
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-338-148 500 mL Glass Bottle
01-338-150 4 L Poly Bottle
01-338-149 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-338-148 Supplier Reagents Holdings Llc Supplier No. C1120300500C
Only null left
Add to Cart
Add to Cart

Iso-Butyl Alcohol, ACS Reagent (2-Methyl-1-Propanol), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 78-83-1
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
InChI Key ZXEKIIBDNHEJCQ-UHFFFAOYSA-N
Synonym Isobutanol, 2-Methyl-1-propanol, 2-Methylpropan-1-ol
IUPAC Name 2-methylpropan-1-ol
SMILES CC(C)CO

Specifications

Melting Point -108°C
Color Colorless
Quantity 500 mL
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.8016
Boiling Point 108°C
Grade ACS
Chemical Name or Material Iso-Butyl Alcohol, ACS Reagent (2-Methyl-1-Propanol)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.