Learn More
Thomas Scientific IsoFreeze Rack, 0 degrees celsius
SDP
Supplier:  Thomas Scientific 1148D57
Encompass
Temperature maintenance racks. Lid included. Consistent thermal protection. One side holds 20 x 1.5/2.0 mL tubes, and the other side holds 20 x 0.5 mL tubes. Tough polycarbonate material.
Catalog No. NC1776817