Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iso-Propyl Alcohol, Purified (2-Propanol; Isopropanol), Reagents

Catalog No. 01337514
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
HDPE Winpak
Poly Bottle
Steel Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-337-514 20 L Steel Pail
01-337-516 1 L Poly Bottle
01-337-513 20 L HDPE Winpak
01-337-512 4 L Poly Bottle
01-337-511 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01-337-514 Supplier Reagents Holdings Llc Supplier No. C212150020E6
Add to Cart
Add to Cart

Iso-Propyl Alcohol, Purified (2-Propanol; Isopropanol), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym Isopropyl Alcohol, 2-Propanol, Propan-2-ol
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point -90°C
Color Colorless
Quantity 20 L
Packaging Steel Pail
Physical Form Liquid
Specific Gravity 0.7808
Boiling Point 82.5°C
Grade Reagent
Chemical Name or Material Iso-Propyl Alcohol, Purified (2-Propanol, Isopropanol)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.