Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Iso-Sensitest Agar (Dehydrated)

Catalog No. OXCM0471B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0471B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0471B Supplier Thermo Scientific™ Supplier No. CM0471B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Perform effortless antimicrobial susceptibility testing using this carefully designed medium to give reproducible results.

Perform effortless antimicrobial susceptibility testing using Thermo Scientific™ Oxoid™ Iso-Sensitest Agar (Dehydrated), a semi-defined medium specifically designed for antimicrobial susceptibility testing (AST). Iso-Sensitest Broth is carefully constructed to give reproducible results. It allows the growth of the majority of microorganisms without the need for further supplementation.

Iso-Sensitest Agar has been designed to overcome objections made about Mueller-Hinton Media by various workers 1,2,3,4,5,6,7

These objections were:

  1. Different MIC values in the broth and agar versions of the medium.
  2. Agar versions showing antagonistic effects towards tetracycline.
  3. High levels of sulphonamide and trimethoprim antagonists.
  4. Poor reproducibility with different manufacturers' peptones.
  5. Poor growth-supporting ability for streptococci and variable growth rates with gram-positive organisms generally

Some mutant strains which are totally dependent on thymine and thymidine for their growth will, however, not grow in Iso-Sensitest Agar, which has these two compounds at very low levels, as they are the naturally occurring antagonists of trimethoprim. Care must be taken to recognize these strains8,9,10.

Iso-Sensitest Agar was developed from Oxoid™ Sensitest Agar (CM0409) which has been used successfully in several centers throughout the world11,12,13. The role of metal ions as antagonists to certain antibiotics has been described by many workers14,15,16,17,18,19. A stabilized mineral content in sensitivity test media is, therefore, important.

Reller, Schoenknecht, Kenny & Sherris3 demonstrated the contribution of cations provided in media by ordinary agars. A considerable difference in mineral content between the agar and broth media was shown. Only by using an agar which is specially processed to remove the free anions and cations can a stabilized mineral content be obtained.

Iso-Sensitest Agar has stability in mineral content and the ability to produce optimum antimicrobial zones of inhibition. The amino-nitrogen base of acid-hydrolyzed casein and special peptones has been supplemented with defined growth factors. Careful preparation of the nutrients ensures that trimethoprim and sulphonamide antagonists are at very low levels.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Iso-Sensitest Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 15.9L medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.