Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoamyl 4-(Dimethylamino)benzoate 98.0+%, TCI America™
SDP

Catalog No. I1013100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I1013100G 100 g
I101325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I1013100G Supplier TCI America Supplier No. I1013100G

Chemical Identifiers

CAS 21245-01-2
Molecular Formula C14H21NO2
Molecular Weight (g/mol) 235.327
MDL Number MFCD00059356
InChI Key OFSAUHSCHWRZKM-UHFFFAOYSA-N
Synonym isoamyl 4-dimethylamino benzoate, spectraban, 4-dimethylaminobenzoic acid isoamyl ester, 3-methylbutyl-4-dimethylaminobenzoate, isoamyl para-n,n-dimethylaminobenzoate, 3-methylbutyl 4-dimethylamino benzoate, benzoic acid, 4-dimethylamino-, 3-methylbutyl ester, 3-methylbutyl4-dimethylaminobenzoate, dsstox_cid_26580, dsstox_rid_81739
PubChem CID 88836
IUPAC Name 3-methylbutyl 4-(dimethylamino)benzoate
SMILES CC(C)CCOC(=O)C1=CC=C(C=C1)N(C)C

Specifications

Melting Point 37°C
Color White-Yellow
Boiling Point 155°C
Quantity 100 g
Formula Weight 235.33
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material Isoamyl 4-(Dimethylamino)benzoate
EINECSNumber (3)-1436
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.