Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoamyl 4-Methoxycinnamate 95.0+%, TCI America™
SDP

Catalog No. I081525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I081525G 25 g
I08155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I081525G Supplier TCI America Supplier No. I081525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 71617-10-2
Molecular Formula C15H20O3
Molecular Weight (g/mol) 248.322
MDL Number MFCD00583856
InChI Key UBNYRXMKIIGMKK-RMKNXTFCSA-N
Synonym 4-Methoxycinnamic Acid Isoamyl Ester, Isopentyl 4-Methoxycinnamate, 4-Methoxycinnamic Acid Isopentyl Ester
PubChem CID 1549789
IUPAC Name 3-methylbutyl (E)-3-(4-methoxyphenyl)prop-2-enoate
SMILES CC(C)CCOC(=O)C=CC1=CC=C(C=C1)OC

Specifications

Color Yellow
Boiling Point 158°C
Quantity 25 g
Formula Weight 248.32
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Isoamyl 4-Methoxycinnamate
RTECSNumber UD3393500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.