Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutyl 4-Hydroxybenzoate 99.0+%, TCI America™
SDP

Catalog No. H0493100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0493100G 100 g
H049325G 25 g
H0493500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0493100G Supplier TCI America Supplier No. H0493100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2-3-4247
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00020167
InChI Key XPJVKCRENWUEJH-UHFFFAOYSA-N
Synonym isobutyl 4-hydroxybenzoate, isobutylparaben, isobutyl p-hydroxybenzoate, isobutyl paraben, isobutyl-4-hydroxybenzoate, isobutyl parahydroxybenzoate, 4-hydroxybenzoic acid isobutyl ester, benzoic acid, 4-hydroxy-, 2-methylpropyl ester, p-oxybenzoesaureisobutylester, unii-0qqj25x58g
PubChem CID 20240
IUPAC Name 2-methylpropyl 4-hydroxybenzoate
SMILES CC(C)COC(=O)C1=CC=C(C=C1)O

Specifications

Melting Point 76°C
Color White
Quantity 100 g
Formula Weight 194.23
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Isobutyl 4-Hydroxybenzoate
EINECSNumber (3)-1585
RTECSNumber DH2247000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.