Learn More
Sigma Organic Chemistry Isobutyl decanoate | 1G | 30673-38-2 | MFCD00059259
Supplier: Sigma Organic Chemistry CDS0005751G

Isobutyl decanoate , 1G
About this Item:
CAS #: 30673-38-2
MDL #: MFCD00059259
Molecular Weight: 228.37
UNSPSC Code: 12352200
Chemical Formula: C14H28O2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.