Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutyl Hexanoate 98.0+%, TCI America™
SDP

Catalog No. H0110500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0110500ML 500 mL
H011025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0110500ML Supplier TCI America Supplier No. H0110500ML

Chemical Identifiers

CAS 105-79-3
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.268
MDL Number MFCD00048870
InChI Key UXUPPWPIGVTVQI-UHFFFAOYSA-N
Synonym isobutyl hexanoate, isobutyl caproate, hexanoic acid, 2-methylpropyl ester, hexanoic acid, isobutyl ester, 2-methyl-1-propyl caproate, n-caproic acid isobutyl ester, isobutyl caproate natural, unii-2a3x4w9gz0, fema no. 2202, iso-butyl n-hexanoate
PubChem CID 7775
ChEBI CHEBI:87421
IUPAC Name 2-methylpropyl hexanoate
SMILES CCCCCC(=O)OCC(C)C

Specifications

Color Colorless
Quantity 500 mL
Formula Weight 172.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyl Hexanoate
EINECSNumber (2)-0782
RTECSNumber MO8389000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.