Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutyl Isovalerate 98.0+%, TCI America™
SDP

Catalog No. I019725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I019725ML 25 mL
I0197500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I019725ML Supplier TCI America Supplier No. I019725ML

Chemical Identifiers

CAS 589-59-3
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.241
MDL Number MFCD00048349
InChI Key KEBDNKNVCHQIJU-UHFFFAOYSA-N
Synonym Isovaleric Acid Isobutyl Ester
PubChem CID 11514
IUPAC Name 2-methylpropyl 3-methylbutanoate
SMILES CC(C)CC(=O)OCC(C)C

Specifications

Color Colorless
Boiling Point 168°C
Quantity 25 mL
UN Number 3272
Formula Weight 158.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyl Isovalerate
EINECSNumber (2)-0776
RTECSNumber NY1506000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.