Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutyl Propionate 98.0+%, TCI America™
SDP

Catalog No. P0506100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0506100ML 100 mL
P050625ML 25 mL
P0506500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P0506100ML Supplier TCI America Supplier No. P0506100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 540-42-1
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00009307
InChI Key FZXRXKLUIMKDEL-UHFFFAOYSA-N
Synonym isobutyl propionate, isobutyl propanoate, 2-methylpropyl propionate, propanoic acid, 2-methylpropyl ester, propionic acid isobutyl ester, propionic acid, isobutyl ester, methyl isobutyl acetate, 2-methyl-1-propyl propanoate, unii-8q382jsy9t, isobutyl propionate natural
PubChem CID 10895
IUPAC Name 2-methylpropyl propanoate
SMILES CCC(=O)OCC(C)C

Specifications

Color Colorless
Boiling Point 138°C
Quantity 100 mL
UN Number 2394
Formula Weight 130.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyl Propionate
EINECSNumber (2)-0774
RTECSNumber UF4930000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.