Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutylamine Hydrochloride 99.0+%, TCI America™
SDP

Catalog No. I009625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I009625G 25 g
I0096500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I009625G Supplier TCI America Supplier No. I009625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5041-09-8
Molecular Formula C4H12ClN
Molecular Weight (g/mol) 109.60
MDL Number MFCD00060199
InChI Key BSMNBEHEFWDHJD-UHFFFAOYSA-N
Synonym 1-Amino-2-methylpropane Hydrochloride, Isobutylammonium Chloride
PubChem CID 12206183
IUPAC Name 2-methylpropan-1-amine;hydrochloride
SMILES CC(C)CN.Cl

Specifications

Melting Point 178°C
Color White
Quantity 25 g
Formula Weight 109.60
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Isobutylamine Hydrochloride
EINECSNumber (2)-0132
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.