Learn More
Sigma Organic Chemistry Isobutyric acid | 500ML | 79-31-2 | MFCD00002658
Supplier: Sigma Organic Chemistry I1754500ML
Isobutyric acid, 500ML
About This Item:
Synonym(s): 2-Methylpropionic acid
Linear Formula: (CH3)2CHCO2H
Molecular Weight: 88.11
CAS Number: 79-31-2
MDL Number: MFCD00002658
UNSPSC Code: 12352100
Purity: 0.99
Boiling Point: 153-154 C (lit.)
Melting Point: -47 C (lit.)
Density: 0.95 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.393 (lit.)
Storage: room temp
EINECS Number: 201-195-7
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.