Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutyric Acid, 99+%

Catalog No. p-150767
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC122520250 25 g Glass bottle
AC122525000 500 g Glass bottle
AC122520010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC122520250 Supplier Thermo Scientific Chemicals Supplier No. 122520250
Add to Cart
This item is not returnable. View return policy

CAS: 79-31-2 | C4H8O2 | 88.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Acylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-31-2
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00002658
InChI Key KQNPFQTWMSNSAP-UHFFFAOYSA-N
Synonym isobutyric acid, isobutanoic acid, 2-methylpropionic acid, propanoic acid, 2-methyl, dimethylacetic acid, isopropylformic acid, iso-butyric acid, isobutyrate, cenex rp b2, acetic acid, dimethyl
PubChem CID 6590
ChEBI CHEBI:16135
IUPAC Name 2-methylpropanoic acid
SMILES CC(C)C(O)=O

Specifications

Melting Point -47.0°C
Boiling Point 153.0°C to 154.0°C
Color Colorless
Assay Percent Range 99+%
Linear Formula (CH3)2CHCO2H
Beilstein 02, 288
Merck Index 15, 5200
Solubility Information Solubility in water: 210g/L (20°C). Other solubilities: miscible with alcohol, chloroform and ether
Formula Weight 88.11
Percent Purity ≥99%
Density 0.9500g/mL
Quantity 25 g
Flash Point 59°C
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.3920 to 1.3940
Specific Gravity 0.95
Chemical Name or Material Isobutyric acid, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-195-7
RTECSNumber NQ4375000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.