Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutyronitrile, 99%, Spectrum™ Chemical
SDP

Catalog No. 18603513 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mL
1 L
200 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-513 200 L Amber Glass Bottle
18-603-511 100 mL Amber Glass Bottle
18-603-512 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-603-513 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. I2199200LTBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

I2199, 78-82-0, C4H7N

Spectrum™ Chemical Isobutyronitrile

Chemical Identifiers

CAS 78-82-0
Molecular Formula C4H7N
Molecular Weight (g/mol) 69.11
InChI Key LRDFRRGEGBBSRN-UHFFFAOYSA-N
IUPAC Name 2-methylpropanenitrile
SMILES CC(C)C#N

Specifications

CAS Max % 100%
Boiling Point 103.9°C/219.2°F
Packaging Amber Glass Bottle
Quantity 200 L
UN Number UN2284
Percent Purity 99%
Grade Reagent
Physical Form Liquid
Chemical Name or Material Isobutyronitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.