Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isochlortetracycline hydrochloride, 97%, can be used as secondary standard, Thermo Scientific™

Catalog No. AC268215000
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC268215000 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC268215000 Supplier Thermo Scientific Chemicals Supplier No. 268215000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89835-80-3
Molecular Formula C22H23ClN2O8·HCl
Synonym ISOCTC

Specifications

Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 500 mg
Formula Weight 515.34
Percent Purity ≥97.0% (on anhydrous substance), ≥80% by HPLC (as is, calculated)
Physical Form Powder
Chemical Name or Material Isochlortetracycline hydrochloride, 97%
Hazard Category LIGHT SENSITIVE
Hazard Statement MOISTURE SENSITIVE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.