Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoconazole nitrate, 99%, Thermo Scientific™

Catalog No. AC458640250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458640250 25 g
AC458640010 1 g
AC458640050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC458640250 Supplier Thermo Scientific Chemicals Supplier No. 458640250
Add to Cart

Chemical Identifiers

CAS 24168-96-5
Molecular Formula C18H15Cl4N3O4
Molecular Weight (g/mol) 479.14
MDL Number MFCD01721647
InChI Key NNGQLSIGRSTLLU-UHFFFAOYNA-N
Synonym isoconazole nitrate, adestan g100, travogyn, fazol, isoconazole; nitric acid, isoconazole nitrate jan, 1-2,4-dichloro-beta-2,6-dichlorobenzyloxy phenethyl imidazole nitrate, 1-2-2,6-dichlorobenzyl oxy-2-2,4-dichlorophenyl ethyl imidazole nitrate
PubChem CID 159968
IUPAC Name 1-[2-(2,4-dichlorophenyl)-2-[(2,6-dichlorophenyl)methoxy]ethyl]imidazole;nitric acid
SMILES O[N+]([O-])=O.ClC1=CC=C(C(CN2C=CN=C2)OCC2=C(Cl)C=CC=C2Cl)C(Cl)=C1

Specifications

Melting Point 181.0°C to 183.0°C
Infrared Spectrum Authentic
Quantity 25 g
Formula Weight 479.14
Percent Purity 99%
Chemical Name or Material Isoconazole nitrate
EINECSNumber 246-051-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.